Thermo Scientific Chemicals

2,4-Lutidine, 98+%, Thermo Scientific Chemicals

Catalog number: B22913.22
100 g, Each
Thermo Scientific Chemicals

2,4-Lutidine, 98+%, Thermo Scientific Chemicals

Catalog number: B22913.22
100 g, Each
Quantity
Catalog number: B22913.22
also known as B22913-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-47-4
IUPAC Name
2,4-dimethylpyridine
Molecular Formula
C7H9N
InChI Key
JYYNAJVZFGKDEQ-UHFFFAOYSA-N
SMILES
CC1=CC=NC(C)=C1
Appearance (Color)
Clear colorless to pale yellow
Comment
Material sourced in UK and US
Form
Liquid
Assay (GC)
≥98.0% (UK-sourced material)
Assay from Supplier's CofA
≥98.0% (US-sourced material)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text