Thermo Scientific Chemicals

2,3,4,5,6-Pentafluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B22922.14
25 g, Each
Thermo Scientific Chemicals

2,3,4,5,6-Pentafluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B22922.14
25 g, Each
Quantity
Catalog number: B22922.14
also known as B22922-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1582-24-7
IUPAC Name
(2,3,4,5,6-pentafluorophenyl)boronic acid
Molecular Formula
C6H2BF5O2
InChI Key
VASOMTXTRMYSKD-UHFFFAOYSA-N
SMILES
OB(O)C1=C(F)C(F)=C(F)C(F)=C1F
Appearance (Color)
White to pale brown
Assay (HPLC)
≥96.0%
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0%

Description

2,3,4,5,6-Pentafluorobenzeneboronic acid is utilized in the palladium-catalyzed Suzuki reaction to synthesize several cross-coupled aromatic compounds, for example bis(pentafluorophenyl)-substituted thiophene and selenophene derivatives. It serves as an intermediate and raw material in pharmaceutical and chemical industry. It is also used in dyes and pigments. It can be used to prepare cyclic boronates of bifunctional compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluorobenzeneboronic acid is utilized in the palladium-catalyzed Suzuki reaction to synthesize several cross-coupled aromatic compounds, for example bis(pentafluorophenyl)-substituted thiophene and selenophene derivatives. It serves as an intermediate and raw material in pharmaceutical and chemical industry. It is also used in dyes and pigments. It can be used to prepare cyclic boronates of bifunctional compounds.

Solubility
Miscible with alcohols, acetonitrile and dichloromethane.

Notes
Moisture Sensitive. Store in cool and dry conditions in well sealed containers. Keep container tightly sealed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text