Thermo Scientific Chemicals

Phenylphosphonic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22939.22
100 g, Each
Thermo Scientific Chemicals

Phenylphosphonic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22939.22
100 g, Each
Quantity
Catalog number: B22939.22
also known as B22939-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1571-33-1
IUPAC Name
phenylphosphonic acid
Molecular Formula
C6H7O3P
InChI Key
QLZHNIAADXEJJP-UHFFFAOYSA-N
SMILES
OP(O)(=O)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms

Description

Phenylphosphonic acid additive used with Noyori's catalyst for oxidation of sulfides to sulfones. Used as additive of unsaturated polyester PU resin. It also can be added into nylon to improve its polymerization degree. It can be used as extreme pressure agent of lube; used as catalyst of carboxylic acid & alcohol reaction; it can be used as fire retarding treatment of fiber used asganic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylphosphonic acid additive used with Noyori′s catalyst for oxidation of sulfides to sulfones. Used as additive of unsaturated polyester PU resin. It also can be added into nylon to improve its polymerization degree. It can be used as extreme pressure agent of lube; used as catalyst of carboxylic acid & alcohol reaction; it can be used as fire retarding treatment of fiber used asganic materials.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text