Thermo Scientific Chemicals

2-(Methylthio)ethylamine, 95%, Thermo Scientific Chemicals

Catalog number: B22941.03
1 g, Each
Thermo Scientific Chemicals

2-(Methylthio)ethylamine, 95%, Thermo Scientific Chemicals

Catalog number: B22941.03
1 g, Each
Quantity
Catalog number: B22941.03
also known as B22941-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18542-42-2
IUPAC Name
2-(methylsulfanyl)ethan-1-aminium
Molecular Formula
C3H10NS
InChI Key
CYWGSFFHHMQKET-UHFFFAOYSA-O
SMILES
CSCC[NH3+]
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.4925-1.5010 @ 20?C
Assay (GC)
≥94.0%

Description

2-(Methylthio)ethylamine is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)ethylamine is a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents, carbon dioxide. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text