Thermo Scientific Chemicals

tert-Butyl ethyl ether, 99%, Thermo Scientific Chemicals

Catalog number: B22945.06
5 g, Each
Thermo Scientific Chemicals

tert-Butyl ethyl ether, 99%, Thermo Scientific Chemicals

Catalog number: B22945.06
5 g, Each
Quantity
Catalog number: B22945.06
also known as B22945-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
637-92-3
IUPAC Name
2-ethoxy-2-methylpropane
Molecular Formula
C6H14O
InChI Key
NUMQCACRALPSHD-UHFFFAOYSA-N
SMILES
CCOC(C)(C)C
Refractive Index
1.3740-1.3770 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%

Description

tert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.

Solubility
Miscible with alcohol, ethyl ether. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, neoprene, n-butyl and urethane. It may react with air to form peroxides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text