L-(-)-Bornyl acetate, 95%, Thermo Scientific Chemicals
L-(-)-Bornyl acetate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-(-)-Bornyl acetate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number B22957.22
also known as B22957-22
Price (USD)/ Each
55.40
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
55.40
Add to cart
L-(-)-Bornyl acetate, 95%, Thermo Scientific Chemicals
Catalog numberB22957.22
Price (USD)/ Each
55.40
-
Add to cart
Chemical Identifiers
CAS5655-61-8
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4620 - 1.4650 @20?C
Appearance (Color)Clear, colourless
Assay (GC)> 94%
Optical Rotation-40.5? ? 5? (neat)
Formliquid/low melting solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only