alpha-Acetyl-gamma-butyrolactone, 98+%, Thermo Scientific Chemicals
alpha-Acetyl-gamma-butyrolactone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

alpha-Acetyl-gamma-butyrolactone, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number B22967.36
also known as B22967-36
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
134.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
134.00 
Save 20.35 (15%)
Add to cart
alpha-Acetyl-gamma-butyrolactone, 98+%, Thermo Scientific Chemicals
Catalog numberB22967.36
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
134.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
CAS517-23-7
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥98.0%
Appearance (Color)Clear colorless
Refractive Index1.4575-1.4615 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only