Thermo Scientific Chemicals

Julolidine, 98%, Thermo Scientific Chemicals

Catalog number: B22976.03
1 g, Each
Thermo Scientific Chemicals

Julolidine, 98%, Thermo Scientific Chemicals

Catalog number: B22976.03
1 g, Each
Quantity
Catalog number: B22976.03
also known as B22976-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
479-59-4
IUPAC Name
1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene
Molecular Formula
C12H15N
InChI Key
DZFWNZJKBJOGFQ-UHFFFAOYSA-N
SMILES
C1CN2CCCC3=C2C(C1)=CC=C3
Appearance (Color)
White/colorless to pink, yellow or cream
Assay (GC)
≥97.5%
Melting Point (clear melt)
32-39?C
Form
Crystalline or fused solid, lumps or powder

Description

Julolidine used as amine building block and in chemoluminescent dye. Also used in photoconductive materials, chemiluminescence substances, chromogenic substrates in analytical redox reactions, dye intermediates, potential antidepressants and tranquilizers, nonlinear optical materials, high sensitivity photopolymerizable materials, and for improving color stability in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Julolidine used as amine building block and in chemoluminescent dye. Also used in photoconductive materials, chemiluminescence substances, chromogenic substrates in analytical redox reactions, dye intermediates, potential antidepressants and tranquilizers, nonlinear optical materials, high sensitivity photopolymerizable materials, and for improving color stability in photography.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents, carbon dioxide. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text