Julolidine used as amine building block and in chemoluminescent dye. Also used in photoconductive materials, chemiluminescence substances, chromogenic substrates in analytical redox reactions, dye intermediates, potential antidepressants and tranquilizers, nonlinear optical materials, high sensitivity photopolymerizable materials, and for improving color stability in photography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Julolidine used as amine building block and in chemoluminescent dye. Also used in photoconductive materials, chemiluminescence substances, chromogenic substrates in analytical redox reactions, dye intermediates, potential antidepressants and tranquilizers, nonlinear optical materials, high sensitivity photopolymerizable materials, and for improving color stability in photography.
Solubility
Soluble in toluene. Insoluble in water.
Notes
Air sensitive. Store away from air, oxidizing agents, carbon dioxide. Store under dry inert gas.
RUO – Research Use Only
General References:
- Michael D. Zammit; Thomas P. Davis and Gary D. Willett. Visible Light Pulsed-Laser Polymerization at 532 nm Employing a Julolidine Dye Photosensitizer Initiation System. Macromolecules. 1997, 30 (19), 5655-5659.
- Hyunbong Choi; Jae Kwan Lee; Kyu Ho Song; Kihyung Song; Sang Ook Kang; Jaejung Ko. Synthesis of new julolidine dyes having bithiophene derivatives for solar cell. Tetrahedron. 2007, 63 7, 1553-1559.