Thermo Scientific Chemicals

3-Methoxy-4-methylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: B22982.14
25 g, Each
Thermo Scientific Chemicals

3-Methoxy-4-methylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: B22982.14
25 g, Each
Quantity
Catalog number: B22982.14
also known as B22982-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7151-68-0
IUPAC Name
3-methoxy-4-methylbenzoic acid
Molecular Formula
C9H10O3
InChI Key
CEAVPXDEPGAVDA-UHFFFAOYSA-N
SMILES
COC1=CC(=CC=C1C)C(O)=O
Appearance (Color)
White to cream
Form
Crystalline powder
Assay (Aqueous acid-base Titration)
>98.5 to <101.5%
Assay (Silylated GC)
>98.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text