Ethyl 3-phenylglycidate, cis + trans, 90+%, Thermo Scientific Chemicals
Ethyl 3-phenylglycidate, cis + trans, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 3-phenylglycidate, cis + trans, 90+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
Catalog number B22990.AK
also known as B22990-AK
Price (USD)/ Each
82.20
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
82.20
Add to cart
Ethyl 3-phenylglycidate, cis + trans, 90+%, Thermo Scientific Chemicals
Catalog numberB22990.AK
Price (USD)/ Each
82.20
-
Add to cart
Chemical Identifiers
CAS121-39-1
IUPAC Nameethyl 3-phenyloxirane-2-carboxylate
Molecular FormulaC11H12O3
InChI KeyGOMAKLPNAAZVCJ-UHFFFAOYNA-N
SMILESCCOC(=O)C1OC1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥90.0%
Refractive Index1.4785-1.5585 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only