Methyl 2-nitrobenzoate, 98+%, Thermo Scientific Chemicals
Methyl 2-nitrobenzoate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 2-nitrobenzoate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B23009.14
also known as B23009-14
Price (USD)/ Each
110.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
110.00
Add to cart
Methyl 2-nitrobenzoate, 98+%, Thermo Scientific Chemicals
Catalog numberB23009.14
Price (USD)/ Each
110.00
-
Add to cart
Chemical Identifiers
CAS606-27-9
IUPAC Namemethyl 2-nitrobenzoate
Molecular FormulaC8H7NO4
InChI KeyAOXPHVNMBPFOFS-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.5325-1.5365 @ 20?C
FormLiquid
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only