Sebaconitrile, 95%, Thermo Scientific Chemicals
Sebaconitrile, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sebaconitrile, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B23010.06
also known as B23010-06
Price (USD)/ Each
26.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
26.00
Add to cart
Sebaconitrile, 95%, Thermo Scientific Chemicals
Catalog numberB23010.06
Price (USD)/ Each
26.00
-
Add to cart
Chemical Identifiers
CAS1871-96-1
IUPAC Namedecanedinitrile
Molecular FormulaC10H16N2
InChI KeyDFJYZCUIKPGCSG-UHFFFAOYSA-N
SMILESN#CCCCCCCCCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥94.0%
Refractive Index1.4465-1.4500 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only