Thermo Scientific Chemicals

Zinc gluconate hydrate, 97%, Thermo Scientific Chemicals

Catalog number: B23022.22
100 g, Each
Thermo Scientific Chemicals

Zinc gluconate hydrate, 97%, Thermo Scientific Chemicals

Catalog number: B23022.22
100 g, Each
Quantity
Catalog number: B23022.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4468-02-4
IUPAC Name
zinc(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
Molecular Formula
C12H22O14Zn
InChI Key
WHMDKBIGKVEYHS-IYEMJOQQSA-L
SMILES
[Zn++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
Appearance (Color)
White
Water Content (Karl Fischer Titration)
3.80-10.60% (1-3 waters)
Trace Metal
(Cadmium, Cd) : ≤ 5ppm
Trace Metal
(Lead, Pb) : ≤ 0.8ppm
Chloride content
≤ 0.05%

Description

Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.

Solubility
Soluble in water 100g/ l.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text