Zinc gluconate hydrate, 97%, Thermo Scientific Chemicals
Zinc gluconate hydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc gluconate hydrate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number B23022.36
also known as B23022-36
Price (USD)/ Each
135.65
Online exclusive
151.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
135.65
Online exclusive
151.00 
Save 15.35 (10%)
Add to cart
Zinc gluconate hydrate, 97%, Thermo Scientific Chemicals
Catalog numberB23022.36
Price (USD)/ Each
135.65
Online exclusive
151.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS4468-02-4
IUPAC Namezinc(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
Molecular FormulaC12H22O14Zn
InChI KeyWHMDKBIGKVEYHS-IYEMJOQQSA-L
SMILES[Zn++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Water Content (Karl Fischer Titration)3.80-10.60% (1-3 waters)
Trace Metal(Cadmium, Cd) : ≤ 5ppm
Trace Metal(Lead, Pb) : ≤ 0.8ppm
Chloride content≤ 0.05%
View more
Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.

Solubility
Soluble in water 100g/ l.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Sunil Sazawal; Margaret Bentley; Robert E. Black; Pratibha Dhingra; Sherly George; Maharaj K. Bhan. Effect of Zinc Supplementation on Observed Activity in Low Socioeconomic Indian Preschool Children. Pediatrics. 1996, 98 (1), 1132 -1137.
  2. L Saghaie; GH Houshfar; M Neishabor. Synthesis and Determination of Partition Coefficients of Zinc Complexes with Clinical Potential application. IJPR. 2006, 5 (3), 179-189.