Hexanoyl chloride has been used in the synthesis of (±)-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one, 14-methyl-1-octadecene. It is used as crystal liquid, pharmaceutical and organic synthesis material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hexanoyl chloride has been used in the synthesis of (±)-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one, 14-methyl-1-octadecene. It is used as crystal liquid, pharmaceutical and organic synthesis material.
Solubility
Soluble in ether, chloroform.
Notes
Moisture sensitive. Store under inert gas. Incompatible with oxidizing agents, water/ moisture.
RUO – Research Use Only
Aamer Saeed; Hummera Rafique; Muhammad Arshad. Total synthesis and antibacterial screening of ( ± )-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one. Journal of Asian Natural Products Research. 2013, 15 (10), 1112-1122.
Quanjing Zhu; Xiaofeng Chu; Zhaoyan Zhang; Wei-Lin Dai; Kangnian Fan. Effect of the tungsten precursor on the high activity of the WO3/ZrO2 catalyst in the oxidative lactonization of 1,2-benzenedimethanol. Molecules. 2013, 18, (5), 5201-5208.