Thermo Scientific Chemicals

Hexamethylcyclotrisiloxane, 97%, Thermo Scientific Chemicals

Catalog number: B23028.14
25 g, Each
Thermo Scientific Chemicals

Hexamethylcyclotrisiloxane, 97%, Thermo Scientific Chemicals

Catalog number: B23028.14
25 g, Each
Quantity
Catalog number: B23028.14
also known as B23028-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
541-05-9
IUPAC Name
hexamethyl-1,3,5,2,4,6-trioxatrisilinane
Molecular Formula
C6H18O3Si3
InChI Key
HTDJPCNNEPUOOQ-UHFFFAOYSA-N
SMILES
C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O1
Appearance (Color)
White
Assay (GC)
≥96.0%
Melting Point (clear melt)
59.0-69.0?C
Form
Crystals or powder or crystalline powder or lumps
Identification (FTIR)
Conforms

Description

Hexamethylcyclotrisiloxane is used in the preparation of graft polymers and block polymers. It is also a useful biochemical for proteomics research and also acts as an intermediate in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethylcyclotrisiloxane is used in the preparation of graft polymers and block polymers. It is also a useful biochemical for proteomics research and also acts as an intermediate in organic reactions.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible in strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text