Hexamethylcyclotrisiloxane, 97%, Thermo Scientific Chemicals
Hexamethylcyclotrisiloxane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hexamethylcyclotrisiloxane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number B23028.22
also known as B23028-22
Price (USD)/ Each
75.65
Online exclusive
83.90 
Save 8.25 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
75.65
Online exclusive
83.90 
Save 8.25 (10%)
Add to cart
Hexamethylcyclotrisiloxane, 97%, Thermo Scientific Chemicals
Catalog numberB23028.22
Price (USD)/ Each
75.65
Online exclusive
83.90 
Save 8.25 (10%)
-
Add to cart
Chemical Identifiers
CAS541-05-9
IUPAC Namehexamethyl-1,3,5,2,4,6-trioxatrisilinane
Molecular FormulaC6H18O3Si3
InChI KeyHTDJPCNNEPUOOQ-UHFFFAOYSA-N
SMILESC[Si]1(C)O[Si](C)(C)O[Si](C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥96.0%
Melting Point (clear melt)59.0-69.0?C
FormCrystals or powder or crystalline powder or lumps
Identification (FTIR)Conforms
Hexamethylcyclotrisiloxane is used in the preparation of graft polymers and block polymers. It is also a useful biochemical for proteomics research and also acts as an intermediate in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethylcyclotrisiloxane is used in the preparation of graft polymers and block polymers. It is also a useful biochemical for proteomics research and also acts as an intermediate in organic reactions.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible in strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Surita, S. C.; Tansel, B. Evaluation of a Full-Scale Water-Based Scrubber for Removing Siloxanes from Digester Gas: A Case Study. Water Environ. Res. 2015, 87 (5), 444-449.
  2. Montanari, T.; Finocchio, E.; Bozzano, I.; Garuti, G.; Giordano, A.; Pistarino, C.; Busca, G. Purification of landfill biogases from siloxanes by adsorption: A study of silica and 13X zeolite adsorbents on hexamethylcyclotrisiloxane separation. Chem. Eng. J. 2010, 165 (3), 859-863.