2,4-Dinitrobenzenesulfonyl chloride was used to protect primary amines. It was used as starting reagent in the synthesis of tert-butyl 2-[(2,4-dinitrophenyl) sulfonyl]amino acetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Dinitrobenzenesulfonyl chloride was used to protect primary amines. It was used as starting reagent in the synthesis of tert-butyl 2-[(2,4-dinitrophenyl) sulfonyl]amino acetate.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents, bases.
RUO – Research Use Only
Vishwanath Gaitonde & Steven J. Sucheck. Synthesis of β-Glycosyl Amides from N-Glycosyl Dinitrobenzenesulfonamides. Journal of Carbohydrate Chemistry. 2012, 31 (4-6), 1478-1484.
ElRachel J. Ball; Philip S. Green; Nittaya Gale; G. John Langley & Tom Brown. Peptide nucleic acid probes with charged photocleavable mass markers Towards PNA-based MALDI-TOF MS genetic analysis. Artif. DNA PNA XNA. 2010, 1 (1), 27-35.