Thermo Scientific Chemicals

2,4-Dinitrobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B23035.22
100 g, Each
Thermo Scientific Chemicals

2,4-Dinitrobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B23035.22
100 g, Each
Quantity
Catalog number: B23035.22
also known as B23035-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1656-44-6
IUPAC Name
2,4-dinitrobenzene-1-sulfonyl chloride
Molecular Formula
C6H3ClN2O6S
InChI Key
SSFSNKZUKDBPIT-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(C(=C1)[N+]([O-])=O)S(Cl)(=O)=O
Form
Crystals or powder or crystalline powder or granules
Appearance (Color)
Cream to yellow to pale brown
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
100.0-107.0°C

Description

2,4-Dinitrobenzenesulfonyl chloride was used to protect primary amines. It was used as starting reagent in the synthesis of tert-butyl 2-[(2,4-dinitrophenyl) sulfonyl]amino acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dinitrobenzenesulfonyl chloride was used to protect primary amines. It was used as starting reagent in the synthesis of tert-butyl 2-[(2,4-dinitrophenyl) sulfonyl]amino acetate.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text