Thermo Scientific Chemicals

1,5-Diaminopentane, 98%, Thermo Scientific Chemicals

Catalog number: B23039.06
5 g, Each
Thermo Scientific Chemicals

1,5-Diaminopentane, 98%, Thermo Scientific Chemicals

Catalog number: B23039.06
5 g, Each
Quantity
Catalog number: B23039.06
also known as B23039-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
462-94-2
IUPAC Name
pentane-1,5-diamine
Molecular Formula
C5H14N2
InChI Key
VHRGRCVQAFMJIZ-UHFFFAOYSA-N
SMILES
NCCCCCN
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4560-1.4600 @ 20?C
Identification (FTIR)
Conforms

Description

1,5-Diaminopentane is mainly used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diaminopentane is mainly used in organic synthesis.

Solubility
Soluble in water.

Notes
Air sensitive & hygroscopic. Incompatible with acid chlorides, acids, acid anhydrides, strong oxidizing agents, carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text