Thermo Scientific Chemicals

3-Amino-1-propanol, 99%, Thermo Scientific Chemicals

Catalog number: B23041.36
500 g, Each
Thermo Scientific Chemicals

3-Amino-1-propanol, 99%, Thermo Scientific Chemicals

Catalog number: B23041.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B23041.36
also known as B23041-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
156-87-6
IUPAC Name
3-aminopropan-1-ol
Molecular Formula
C3H9NO
InChI Key
WUGQZFFCHPXWKQ-UHFFFAOYSA-N
SMILES
NCCCO
Assay (GC)
≥98.5%
Water Content (Karl Fischer Titration)
≤0.5%
Refractive Index
1.4595-1.4625 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

3-Amino-1-propanol is used in the production of anionic emulsifiers, nonionic polyethylene emulsions. Its salts such as hydrochloride, nitrate, sulfate, phosphate are used for the balanced curing action with selected textile resins. It is used as a starting material in the preparation of beta-lactam antibiotics, humectants for foods and cosmetics. It acts as a corrosion inhibitor and finds application in water treatment, metal treatment and absorption of carbon dioxide gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-1-propanol is used in the production of anionic emulsifiers, nonionic polyethylene emulsions. Its salts such as hydrochloride, nitrate, sulfate, phosphate are used for the balanced curing action with selected textile resins. It is used as a starting material in the preparation of beta-lactam antibiotics, humectants for foods and cosmetics. It acts as a corrosion inhibitor and finds application in water treatment, metal treatment and absorption of carbon dioxide gas.

Solubility
Miscible with water, alcohol, ether, acetone and chloroform.

Notes
Hygroscopic. Incompatible withstrong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text