Thermo Scientific Chemicals

(+/-)-Tetrahydrofurfurylamine, 97%, Thermo Scientific Chemicals

Catalog number: B23068.22
100 g, Each
Thermo Scientific Chemicals

(+/-)-Tetrahydrofurfurylamine, 97%, Thermo Scientific Chemicals

Catalog number: B23068.22
100 g, Each
Quantity
Catalog number: B23068.22
also known as B23068-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4795-29-3
IUPAC Name
1-(oxolan-2-yl)methanamine
Molecular Formula
C5H11NO
InChI Key
YNOGYQAEJGADFJ-UHFFFAOYNA-N
SMILES
NCC1CCCO1
Appearance (Color)
Clear colorless to dark yellow or yellow/green
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4525-1.4575 @ 20?C

Description

Tetrahydrofurfurylamine was used to study specific interactions between unlike molecules by calculating the molar excess enthalpies. It can be used to produce N-tetrahydrofurfuryl-phthalimide with phthalic acid anhydride by heating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrahydrofurfurylamine was used to study specific interactions between unlike molecules by calculating the molar excess enthalpies. It can be used to produce N-tetrahydrofurfuryl-phthalimide with phthalic acid anhydride by heating.

Solubility
Miscible with water.

Notes
Air sensitive & hygroscopic. Store away from oxidizing agents, water/ moisture. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text