Thermo Scientific Chemicals

Thiophene-2-boronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23071.14
25 g, Each
Thermo Scientific Chemicals

Thiophene-2-boronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23071.14
25 g, Each
Quantity
Catalog number: B23071.14
also known as B23071-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6165-68-0
IUPAC Name
(thiophen-2-yl)boronic acid
Molecular Formula
C4H5BO2S
InChI Key
ARYHTUPFQTUBBG-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CS1
Appearance (Color)
White to cream
Form
Powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Identification (FTIR)
Conforms

Description

Thiophene-2-boronic acid is a reagent used for Palladium-catalyzed Suzuki-Miyaura cross-couplings, Copper-catalyzed nitration reactions, Chain-growth catalyst transfer polycondensation of conjugated alternating copolymer, Ferric perchlorate-promoted reaction of fullerene to give fullerenyl boronic esters. It is also used in the preparation of Amino pyridine-based inhibitors of mitotic kinase Nek2 with potential antipoliferative effects in cancer tumors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2-boronic acid is a reagent used for Palladium-catalyzed Suzuki-Miyaura cross-couplings, Copper-catalyzed nitration reactions, Chain-growth catalyst transfer polycondensation of conjugated alternating copolymer, Ferric perchlorate-promoted reaction of fullerene to give fullerenyl boronic esters. It is also used in the preparation of Amino pyridine-based inhibitors of mitotic kinase Nek2 with potential antipoliferative effects in cancer tumors.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text