Thermo Scientific Chemicals

Creatinine, 98%, Thermo Scientific Chemicals

Catalog number: B23097.09
10 g, Each
Thermo Scientific Chemicals

Creatinine, 98%, Thermo Scientific Chemicals

Catalog number: B23097.09
10 g, Each
Quantity
Catalog number: B23097.09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60-27-5
IUPAC Name
2-amino-1-methyl-4,5-dihydro-1H-imidazol-4-one
Molecular Formula
C4H7N3O
InChI Key
DDRJAANPRJIHGJ-UHFFFAOYSA-N
SMILES
CN1CC(=O)N=C1N
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms

Description

Creatinine levels in blood and urine may be used to calculate the creatinine clearance (CrCl), which reflects the glomerular filtration rate (GFR), an important clinical index of renal function. It is a substrate for creatininase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Creatinine levels in blood and urine may be used to calculate the creatinine clearance (CrCl), which reflects the glomerular filtration rate (GFR), an important clinical index of renal function. It is a substrate for creatininase.

Solubility
Soluble in water (90 mg/ml at 20°C), alcohol (slightly), and ethanol (slightly). Insoluble in acetone, and chloroform.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text