Thermo Scientific Chemicals

(1R,3S)-(+)-Camphoric acid, 98+%, Thermo Scientific Chemicals

Catalog number: B23106.14
25 g, Each
Thermo Scientific Chemicals

(1R,3S)-(+)-Camphoric acid, 98+%, Thermo Scientific Chemicals

Catalog number: B23106.14
25 g, Each
Quantity
Catalog number: B23106.14
also known as B23106-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-83-4
IUPAC Name
(1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate
Molecular Formula
C10H14O4
InChI Key
LSPHULWDVZXLIL-LDWIPMOCSA-L
SMILES
CC1(C)[C@H](CC[C@@]1(C)C([O-])=O)C([O-])=O
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Melting Point (clear melt)
184.5-190.5?C
Optical Rotation
46 ± 2? (c=10 in ethanol)
Appearance (Color)
White
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text