Thermo Scientific Chemicals

Methyltrichlorosilane, 97%, Thermo Scientific Chemicals

Catalog number: B23107.36
500 g, Each
Thermo Scientific Chemicals

Methyltrichlorosilane, 97%, Thermo Scientific Chemicals

Catalog number: B23107.36
500 g, Each
Quantity
Catalog number: B23107.36
also known as B23107-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-79-6
IUPAC Name
trichloro(methyl)silane
Molecular Formula
CH3Cl3Si
InChI Key
JLUFWMXJHAVVNN-UHFFFAOYSA-N
SMILES
C[Si](Cl)(Cl)Cl
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.4090-1.4140 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid

Description

Methyltrichlorosilane is used in production of methyl silicone resins, its vapor reacts with water on surfaces to give a thin layer of methylpolysiloxane which make it a water-repellent film. A combination of methyltrichlorosilane and sodium iodide can be used to cleave carbon-oxygen bonds such as methyl ethers. It is used as a precursor for forming various cross-linked siloxane polymers. Trichloromethylsilane is the starting material for the production of pure silicon for manufacture of semiconductors and optical fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltrichlorosilane is used in production of methyl silicone resins, its vapor reacts with water on surfaces to give a thin layer of methylpolysiloxane which make it a water-repellent film. A combination of methyltrichlorosilane and sodium iodide can be used to cleave carbon-oxygen bonds such as methyl ethers. It is used as a precursor for forming various cross-linked siloxane polymers. Trichloromethylsilane is the starting material for the production of pure silicon for manufacture of semiconductors and optical fibers.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text