Thermo Scientific™

1-Ethynylcyclopentanol, 98+%, Thermo Scientific Chemicals

Catalog number: B23109.06
5 g, Each
Thermo Scientific™

1-Ethynylcyclopentanol, 98+%, Thermo Scientific Chemicals

Catalog number: B23109.06
5 g, Each
Quantity
Catalog number: B23109.06
also known as B23109-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17356-19-3
IUPAC Name
1-ethynylcyclopentan-1-ol
Molecular Formula
C7H10O
InChI Key
LQMDOONLLAJAPZ-UHFFFAOYSA-N
SMILES
OC1(CCCC1)C#C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.4720-1.4760 @ 20?C

Description

1-Ethynylcyclopentanol is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethynylcyclopentanol is used in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text