Thermo Scientific Chemicals

Diphenylcarbamyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B23116.14
25 g, Each
Thermo Scientific Chemicals

Diphenylcarbamyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B23116.14
25 g, Each
Quantity
Catalog number: B23116.14
also known as B23116-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
83-01-2
IUPAC Name
N,N-diphenylcarbamoyl chloride
Molecular Formula
C13H10ClNO
InChI Key
XNBKKRFABABBPM-UHFFFAOYSA-N
SMILES
ClC(=O)N(C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Assay (GC)
≥97.5%
Melting Point (clear melt)
82.0-88.0?C

Description

Diphenylcarbamoyl chloride was used to isolate peptido-keratan sulphate fragments from bovine intervertebral discs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylcarbamoyl chloride was used to isolate peptido-keratan sulphate fragments from bovine intervertebral discs.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text