2-Chloro-4-fluorophenylacetonitrile, 97%, Thermo Scientific Chemicals
2-Chloro-4-fluorophenylacetonitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-4-fluorophenylacetonitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B23132.06
also known as B23132-06
Price (USD)/ Each
35.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
35.80
Add to cart
2-Chloro-4-fluorophenylacetonitrile, 97%, Thermo Scientific Chemicals
Catalog numberB23132.06
Price (USD)/ Each
35.80
-
Add to cart
Chemical Identifiers
CAS75279-56-0
IUPAC Name2-(2-chloro-4-fluorophenyl)acetonitrile
Molecular FormulaC8H5ClFN
InChI KeyGSMCLMKFBYLWRP-UHFFFAOYSA-N
SMILESFC1=CC=C(CC#N)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)63.0-73.0?C
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only