Thermo Scientific Chemicals

Diethoxymethane, 99%, stab. with 100ppm BHT, Thermo Scientific Chemicals

Catalog number: B23134.AP
500 mL, Each
Thermo Scientific Chemicals

Diethoxymethane, 99%, stab. with 100ppm BHT, Thermo Scientific Chemicals

Catalog number: B23134.AP
500 mL, Each
Quantity
Catalog number: B23134.AP
also known as B23134-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
462-95-3
IUPAC Name
(ethoxymethoxy)ethane
Molecular Formula
C5H12O2
InChI Key
KLKFAASOGCDTDT-UHFFFAOYSA-N
SMILES
CCOCOCC
Form
Liquid
Appearance (Color)
Clear colourless
Assay (GC)
> 98.5%
Refractive Index
1.3720 - 1.3760 @20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text