Thermo Scientific Chemicals

Terephthalonitrile, 98%, Thermo Scientific Chemicals

Catalog number: B23137.14
25 g, Each
Thermo Scientific Chemicals

Terephthalonitrile, 98%, Thermo Scientific Chemicals

Catalog number: B23137.14
25 g, Each
Quantity
Catalog number: B23137.14
also known as B23137-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
623-26-7
IUPAC Name
benzene-1,4-dicarbonitrile
Molecular Formula
C8H4N2
InChI Key
BHXFKXOIODIUJO-UHFFFAOYSA-N
SMILES
N#CC1=CC=C(C=C1)C#N
Appearance (Color)
White to pale yellow or pale cream
Form
Powder
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
222-228?C

Description

1,4-dicyanobenzene as a new internal standard in utilizing solid-state infrared intensity methods for the determination of thiocyanate bonding. Used as photosensitizer in [2+2] cycloadditions, nitrile building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-dicyanobenzene as a new internal standard in utilizing solid-state infrared intensity methods for the determination of thiocyanate bonding. Used as photosensitizer in [2+2] cycloadditions, nitrile building block.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text