4-(4-Fluorobenzoyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals
4-(4-Fluorobenzoyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(4-Fluorobenzoyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number B23142.06
also known as B23142-06
Price (USD)/ Each
231.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
231.00
Add to cart
4-(4-Fluorobenzoyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberB23142.06
Price (USD)/ Each
231.00
-
Add to cart
Chemical Identifiers
CAS25519-78-2
IUPAC Namehydrogen 4-(4-fluorobenzoyl)piperidine chloride
Molecular FormulaC12H15ClFNO
InChI KeyGPKDBZQZPNOBGM-UHFFFAOYSA-N
SMILES[H+].[Cl-].FC1=CC=C(C=C1)C(=O)C1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)223.0-229.0?C
FormPowder
Assay (Titration ex Chloride)≥97.5%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only