Thermo Scientific Chemicals

2-Methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B23154.06
5 g, Each
Thermo Scientific Chemicals

2-Methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B23154.06
5 g, Each
Quantity
Catalog number: B23154.06
also known as B23154-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16419-60-6
IUPAC Name
(2-methylphenyl)boronic acid
Molecular Formula
C7H9BO2
InChI Key
NSJVYHOPHZMZPN-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1B(O)O
Assay (Aqueous acid-base Titration)
≥97.5%
Assay (HPLC)
≥97.5%
Proton NMR
Conforms to structure
Appearance (Color)
White to cream
Comment
May contain varying amounts of anhydride

Description

2-Methylbenzeneboronic acid is a reagent used for Pd-catalyzed arylation using Suzuki-Miyaura cross-coupling in water, Ruthenium catalyzed direct arylation reactions, Ligand-free copper-catalyzed coupling reactions, Rhodium-catalyzed asymmetric 1,4-addition reactions. Reagent used in Preparation of chiral monophosphorus ligands in asymmetric Suzuki-Miyaura coupling reaction, Human farnesyl pyrophosphate synthase inhibitors as antitumor agents for multiple myeloma cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzeneboronic acid is a reagent used for Pd-catalyzed arylation using Suzuki-Miyaura cross-coupling in water, Ruthenium catalyzed direct arylation reactions, Ligand-free copper-catalyzed coupling reactions, Rhodium-catalyzed asymmetric 1,4-addition reactions. Reagent used in Preparation of chiral monophosphorus ligands in asymmetric Suzuki-Miyaura coupling reaction, Human farnesyl pyrophosphate synthase inhibitors as antitumor agents for multiple myeloma cells

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text