Thermo Scientific Chemicals

Tetraethylammonium fluoride hydrate, 97%, Thermo Scientific Chemicals

Catalog number: B23173.14
25 g, Each
Thermo Scientific Chemicals

Tetraethylammonium fluoride hydrate, 97%, Thermo Scientific Chemicals

Catalog number: B23173.14
25 g, Each
Quantity
Catalog number: B23173.14
also known as B23173-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98330-04-2
IUPAC Name
tetraethylazanium fluoride
Molecular Formula
C8H20FN
InChI Key
QSUJAUYJBJRLKV-UHFFFAOYSA-M
SMILES
[F-].CC[N+](CC)(CC)CC
Comment
Specification differs for U.S. and non-U.S. material where indicated
Water Content (Karl Fischer Titration)
≤30% (non-U.S. specification)
Appearance (Color)
White to pale cream
Form
Powder or crystals or crystalline powder or lumpy crystalline powder
Assay (Non-aqueous acid-base Titration)
≥96.0 to <104.0% (dry wt. basis, non-U.S. specification)

Description

Tetraethylammonium fluoride hydrate is used as a catalyst for oxidation, methylation and deuteration reactions of fluorine. It is involved as a reagent for the cleavage of Si-C bonds and also acts as a phase transfer catalyst in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium fluoride hydrate is used as a catalyst for oxidation, methylation and deuteration reactions of fluorine. It is involved as a reagent for the cleavage of Si-C bonds and also acts as a phase transfer catalyst in synthetic chemistry.

Solubility
Soluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text