Thermo Scientific Chemicals

beta-Propiolactone, 97%, Thermo Scientific Chemicals

Catalog number: B23197.06
5 g, Each
Thermo Scientific Chemicals

beta-Propiolactone, 97%, Thermo Scientific Chemicals

Catalog number: B23197.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B23197.06
also known as B23197-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-57-8
IUPAC Name
oxetan-2-one
Molecular Formula
C3H4O2
InChI Key
VEZXCJBBBCKRPI-UHFFFAOYSA-N
SMILES
O=C1CCO1
Form
Liquid
Refractive Index
1.4105-1.4155 @ 20°C (non-U.S. specification)
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0% (non-U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

Beta-Propiolactone is widely used as a sterilizing and sporicidal agent, which is used to sterilize blood plasma, vaccines, tissue grafts, surgical instruments and enzymes. It finds an application in the manufacturing of acrylic acid and its esters. It is utilized as an intermediate in the synthesis of synthetic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Beta-Propiolactone is widely used as a sterilizing and sporicidal agent, which is used to sterilize blood plasma, vaccines, tissue grafts, surgical instruments and enzymes. It finds an application in the manufacturing of acrylic acid and its esters. It is utilized as an intermediate in the synthesis of synthetic compounds.

Solubility
Miscible with water and organic solvents.

Notes
Flammable material. Should be stored in a separate safety storage cabinet or room. Keep away from heat and sources of ignition.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text