2-(Amino methyl)pyridine is a synthesis reagent used in the synthesis of PPD analogues. It is also used in the preparation of 5-nitro-2-furancarboxylamides displaying potent tryoanocidal activity against trypanosomiasis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Amino methyl)pyridine is a synthesis reagent used in the synthesis of PPD analogues. It is also used in the preparation of 5-nitro-2-furancarboxylamides displaying potent tryoanocidal activity against trypanosomiasis.
Solubility
Soluble in water (1000 mg/ml at 20°C), and ethanol.
Notes
Air sensitive. Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, acids, air.
RUO – Research Use Only
Francesco Fringuelli; Ferdinando Pizzo; Simone Tortoioli; and Luigi Vaccaro. Solvent-Free Al(OTf)3-Catalyzed Aminolysis of 1,2-Epoxides by 2-Picolylamine: A Key Step in the Synthesis of Ionic Liquids. J. Org. Chem. 2004, 69 (22), 7745-7747.
GJ Sutton. Some Studies in Inorganic Complexes. VI. Nickel(II) with 2-Picolylamine. Australian Journal of Chemistry. 1960, 13 (1), 74 - 79.