Thermo Scientific Chemicals

2-(Aminomethyl)pyridine, 98+%, Thermo Scientific Chemicals

Catalog number: B23208.22
100 g, Each
Thermo Scientific Chemicals

2-(Aminomethyl)pyridine, 98+%, Thermo Scientific Chemicals

Catalog number: B23208.22
100 g, Each
Quantity
Catalog number: B23208.22
also known as B23208-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3731-51-9
IUPAC Name
1-(pyridin-2-yl)methanamine
Molecular Formula
C6H8N2
InChI Key
WOXFMYVTSLAQMO-UHFFFAOYSA-N
SMILES
NCC1=CC=CC=N1
Appearance (Color)
Clear pale yellow to yellow
Form
Liquid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
1.5435-1.5475 @ 20?C

Description

2-(Amino methyl)pyridine is a synthesis reagent used in the synthesis of PPD analogues. It is also used in the preparation of 5-nitro-2-furancarboxylamides displaying potent tryoanocidal activity against trypanosomiasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Amino methyl)pyridine is a synthesis reagent used in the synthesis of PPD analogues. It is also used in the preparation of 5-nitro-2-furancarboxylamides displaying potent tryoanocidal activity against trypanosomiasis.

Solubility
Soluble in water (1000 mg/ml at 20°C), and ethanol.

Notes
Air sensitive. Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, acids, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text