Thermo Scientific Chemicals

Ethyl dibromoacetate, 96%, Thermo Scientific Chemicals

Catalog number: B23209.14
25 g, Each
Thermo Scientific Chemicals

Ethyl dibromoacetate, 96%, Thermo Scientific Chemicals

Catalog number: B23209.14
25 g, Each
Quantity
Catalog number: B23209.14
also known as B23209-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
617-33-4
IUPAC Name
ethyl 2,2-dibromoacetate
Molecular Formula
C4H6Br2O2
InChI Key
NIJGVVHCUXNSLL-UHFFFAOYSA-N
SMILES
CCOC(=O)C(Br)Br
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥95.0%
Refractive Index
1.4960-1.5020 @ 20?C

Description

Ethyl dibromoacetate can be used to produce diphenoxy-acetic acid ethyl ester. It is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl dibromoacetate can be used to produce diphenoxy-acetic acid ethyl ester. It is used in organic synthesis.

Solubility
Soluble in hexane, ether, CH2Cl2, acetone, EtOAc, ethanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text