Thermo Scientific Chemicals

Methyl 2-(chlorosulfonyl)benzoate, 94%, Thermo Scientific Chemicals

Catalog number: B23211.06
5 g, Each
Thermo Scientific Chemicals

Methyl 2-(chlorosulfonyl)benzoate, 94%, Thermo Scientific Chemicals

Catalog number: B23211.06
5 g, Each
Quantity
Catalog number: B23211.06
also known as B23211-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26638-43-7
IUPAC Name
methyl 2-(chlorosulfonyl)benzoate
Molecular Formula
C8H7ClO4S
InChI Key
HUNUAFNLLYVTQD-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=C1S(Cl)(=O)=O
Assay (Aqueous acid-base Titration)
>92.5%
Appearance (Color)
White to pale yellow or cream
Form
Powder

Description

Methyl 2-(chlorosulfonyl)benzoate can be used as intermediates in pharmaceuticals, saccharin, dyes, & pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-(chlorosulfonyl)benzoate can be used as intermediates in pharmaceuticals, saccharin, dyes, & pigments.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text