Cobalt(II) acetate, anhydrous, 98+%, Thermo Scientific Chemicals
Cobalt(II) acetate, anhydrous, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) acetate, anhydrous, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B23218.30
also known as B23218-30
Price (USD)/ Each
821.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
967.00 
Save 145.35 (15%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
821.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
967.00 
Save 145.35 (15%)
Add to cart
Cobalt(II) acetate, anhydrous, 98+%, Thermo Scientific Chemicals
Catalog numberB23218.30
Price (USD)/ Each
821.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
967.00 
Save 145.35 (15%)
-
Add to cart
Chemical Identifiers
CAS71-48-7
IUPAC Nameλ²-cobalt(2+) diacetate
Molecular FormulaC4H6CoO4
InChI KeyQAHREYKOYSIQPH-UHFFFAOYSA-L
SMILES[Co++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink to purple
Formpowder
Assay (unspecified)> 98.0% (ex Cobalt)
Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.

Solubility
Soluble in water, alcohol and dilute acids.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Rajabi, F.; Feiz, A.; Luque, R. An Efficient Synthesis of Coumarin Derivatives Using a SBA-15 Supported Cobalt(II) Nanocatalyst. Catal. Lett. 2015, 145 (8), 1621-1625.
  2. Xu, D.; Xie, Y.; Song, Y. J.; Deng, W. Q. A green and facile method toward synthesis of waste paper-derived 3D functional porous graphene via in situ activation of cobalt(II). J. Mater. Chem. A 2015, 3 (31), 16072-16078.