5,7-Dimethoxycoumarin, 98%, Thermo Scientific Chemicals
5,7-Dimethoxycoumarin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5,7-Dimethoxycoumarin, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
0.25 g
1 g
Catalog number B23226.06
also known as B23226-06
Price (USD)/ Each
393.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
393.00
Add to cart
5,7-Dimethoxycoumarin, 98%, Thermo Scientific Chemicals
Catalog numberB23226.06
Price (USD)/ Each
393.00
-
Add to cart
Chemical Identifiers
CAS487-06-9
IUPAC Name5,7-dimethoxy-2H-chromen-2-one
Molecular FormulaC11H10O4
InChI KeyNXJCRELRQHZBQA-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=C2C=CC(=O)OC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Melting Point (clear melt)145.0-151.0?C
Appearance (Color)White to pale cream to pale yellow to pale brown
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only