N-Isopropylbenzylamine, 97%, Thermo Scientific Chemicals
N-Isopropylbenzylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Isopropylbenzylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B23243.22
also known as B23243-22
Price (USD)/ Each
45.65
Online exclusive
50.60 
Save 4.95 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
45.65
Online exclusive
50.60 
Save 4.95 (10%)
Add to cart
N-Isopropylbenzylamine, 97%, Thermo Scientific Chemicals
Catalog numberB23243.22
Price (USD)/ Each
45.65
Online exclusive
50.60 
Save 4.95 (10%)
-
Add to cart
Chemical Identifiers
CAS102-97-6
IUPAC Namebenzyl(propan-2-yl)amine
Molecular FormulaC10H15N
InChI KeyLYBKPDDZTNUNNM-UHFFFAOYSA-N
SMILESCC(C)NCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (GC)≥96.0%
Refractive Index1.4995-1.5045 @ 20?C
Appearance (Color)Clear colorless to yellow
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only