Thermo Scientific Chemicals

2-Ethylhexyl acetate, 99%, Thermo Scientific Chemicals

Catalog number: B23257.AU
1000 mL, Each
Thermo Scientific Chemicals

2-Ethylhexyl acetate, 99%, Thermo Scientific Chemicals

Catalog number: B23257.AU
1000 mL, Each
Quantity
Catalog number: B23257.AU
also known as B23257-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-09-3
IUPAC Name
2-ethylhexyl acetate
Molecular Formula
C10H20O2
InChI Key
WOYWLLHHWAMFCB-UHFFFAOYNA-N
SMILES
CCCCC(CC)COC(C)=O
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Comment
Specification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)
Conforms (non-U.S. specification)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text