Thermo Scientific Chemicals

Methyl isonicotinate, 98%, Thermo Scientific Chemicals

Catalog number: B23263.36
500 g, Each
Thermo Scientific Chemicals

Methyl isonicotinate, 98%, Thermo Scientific Chemicals

Catalog number: B23263.36
500 g, Each
Quantity
Catalog number: B23263.36
also known as B23263-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2459-09-8
IUPAC Name
methyl pyridine-4-carboxylate
Molecular Formula
C7H7NO2
InChI Key
OLXYLDUSSBULGU-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=NC=C1
Form
Liquid
Water Content (Karl Fischer Titration)
≤0.5%
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to orange to brown

Description

Methyl Isonicotinate induces increased walking and take-off behavior in western flower thrips, Frankliniella occidentalis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl Isonicotinate induces increased walking and take-off behavior in western flower thrips, Frankliniella occidentalis.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text