Thermo Scientific Chemicals

Tri(ethylene glycol) di-p-toluenesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: B23279.18
50 g, Each
Thermo Scientific Chemicals

Tri(ethylene glycol) di-p-toluenesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: B23279.18
50 g, Each
Quantity
Catalog number: B23279.18
also known as B23279-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19249-03-7
IUPAC Name
2-(2-{2-[(4-methylbenzenesulfonyl)oxy]ethoxy}ethoxy)ethyl 4-methylbenzene-1-sulfonate
Molecular Formula
C20H26O8S2
InChI Key
KCONMNWPRXAWKK-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOCCOS(=O)(=O)C1=CC=C(C)C=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
76.0-82.0?C

Description

Tri(ethylene glycol) di-p-toluenesulfonate is used in the synthesis of 1-[4-(1,4,7,10-tetraoxa-13-aza-cyclopentadec-13-yl)-phenyl] ethanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri(ethylene glycol) di-p-toluenesulfonate is used in the synthesis of 1-[4-(1,4,7,10-tetraoxa-13-aza-cyclopentadec-13-yl)-phenyl] ethanone.

Solubility
Soluble in ethanol.

Notes
Incompatible with strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text