Thermo Scientific Chemicals

2-Ethyl-2-oxazoline, 99%, Thermo Scientific Chemicals

Catalog number: B23287.22
100 g, Each
Thermo Scientific Chemicals

2-Ethyl-2-oxazoline, 99%, Thermo Scientific Chemicals

Catalog number: B23287.22
100 g, Each
Quantity
Catalog number: B23287.22
also known as B23287-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10431-98-8
IUPAC Name
2-ethyl-4,5-dihydro-1,3-oxazole
Molecular Formula
C5H9NO
InChI Key
NYEZZYQZRQDLEH-UHFFFAOYSA-N
SMILES
CCC1=NCCO1
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.4365-1.4395 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

2-ethyl-2-oxazoline is an organic intermediate and monomer that is used in water-soluble polymers, pharmaceuticals, cosmetics, specialty coatings and adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-ethyl-2-oxazoline is an organic intermediate and monomer that is used in water-soluble polymers, pharmaceuticals, cosmetics, specialty coatings and adhesives.

Solubility
Fully miscible in water.

Notes
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text