Thermo Scientific Chemicals

Potassium tetrachloroaurate(III) hydrate, 99%, Thermo Scientific Chemicals

Catalog number: B23292.06
5 g, Each
Thermo Scientific Chemicals

Potassium tetrachloroaurate(III) hydrate, 99%, Thermo Scientific Chemicals

Catalog number: B23292.06
5 g, Each
Quantity
Catalog number: B23292.06
also known as B23292-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27988-75-6
IUPAC Name
potassium tetrachlorogoldtris(ylium)
Molecular Formula
AuCl4K
InChI Key
IKHCGJWURGSQRT-UHFFFAOYSA-J
SMILES
[K+].Cl[Au+3](Cl)(Cl)Cl
Appearance (Color)
Yellow to orange
Form
Powder
Elemental Analysis
Gold, Au: 50.33-52.12%

Description

Potassium tetrachloroaurate(III) hydrate is used as primary and secondary intermediates. A Key raw material for synthesis of new gold(III) dithiolate complexes for luminescence studies or possible uses as photosensitizers or photocatalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tetrachloroaurate(III) hydrate is used as primary and secondary intermediates. A Key raw material for synthesis of new gold(III) dithiolate complexes for luminescence studies or possible uses as photosensitizers or photocatalysts.

Solubility
Soluble in water, ethanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text