N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B23302.14
also known as B23302-14
Price (USD)/ Each
37.30
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
37.30
Add to cart
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
Catalog numberB23302.14
Price (USD)/ Each
37.30
-
Add to cart
Chemical Identifiers
CAS3710-84-7
IUPAC NameN,N-diethylhydroxylamine
Molecular FormulaC4H11NO
InChI KeyFVCOIAYSJZGECG-UHFFFAOYSA-N
SMILESCCN(O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4180-1.4230 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only