Thermo Scientific Chemicals

Zirconium n-butoxide, 80% w/w in 1-butanol, Thermo Scientific Chemicals

Catalog number: B23317.0B
1000 g, Each
Thermo Scientific Chemicals

Zirconium n-butoxide, 80% w/w in 1-butanol, Thermo Scientific Chemicals

Catalog number: B23317.0B
1000 g, Each
Quantity
Catalog number: B23317.0B
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1071-76-7
IUPAC Name
butan-1-ol zirconium
Molecular Formula
C4H10OZr
InChI Key
SKFIUGUKJUULEM-UHFFFAOYSA-N
SMILES
[Zr].CCCCO
Comment
Assay typically 84 - 87%
Appearance (Color)
Clear, colourless to pale yellow
Comment
Zirconium as ZrO2 26 - 29% w/w
Assay (unspecified)
81 - 90% w/w
Refractive Index
1.4610 - 1.4700 @20?C

Description

Zirconium(IV) butoxide solution used in the preparation of CeO2-stabilized zirconia polycrystals gel that have good applications as protective coatings of steel and ferrous alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zirconium(IV) butoxide solution used in the preparation of CeO2-stabilized zirconia polycrystals gel that have good applications as protective coatings of steel and ferrous alloys.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, acids, bases, halogens, alkali metals, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text